Anti-CDX2 Mouse Monoclonal Antibody [clone: [1C7]]

51-439
10074-732EA 906.92 CAD
10074-732
Anti-CDX2 Mouse Monoclonal Antibody [clone: [1C7]]
CDX2 antibody can be used in immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

Store CDX2 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: CDX2
Clonality: Monoclonal
Clone: [1C7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR