Anti-DARS Mouse Monoclonal Antibody [clone: [2F11]]

51-447
10074-748EA 906.92 CAD
10074-748
Anti-DARS Mouse Monoclonal Antibody [clone: [2F11]]
DARS antibody can be used in Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

Store DARS antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: DARS
Clonality: Monoclonal
Clone: [2F11]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR