Anti-DCN Mouse Monoclonal Antibody [clone: [3H4-1F4]]

51-449
10076-944EA 906.92 CAD
10076-944
Anti-DCN Mouse Monoclonal Antibody [clone: [3H4-1F4]]
Decorin antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Store Decorin antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: DCN
Clonality: Monoclonal
Clone: [3H4-1F4]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR