Anti-TSC22D3 Mouse Monoclonal Antibody [clone: [3A5]]

Supplier: ProSci

51-450
10076-946EA 1124.21 CAD
10076-946
Anti-TSC22D3 Mouse Monoclonal Antibody [clone: [3A5]]
Antibodies
TSC22D3 antibody can be used in Western Blot starting at 1 - 2 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TSC22D3
Clonality: Monoclonal
Clone: [3A5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR