Anti-GPR3 Mouse Monoclonal Antibody [clone: [3B4-G3]]

51-460
10076-966EA 906.92 CAD
10076-966
Anti-GPR3 Mouse Monoclonal Antibody [clone: [3B4-G3]]
GPR3 antibody can be used in Western Blot starting at 2 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Store GPR3 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: GPR3
Clonality: Monoclonal
Clone: [3B4-G3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR