Anti-CK81 Mouse Monoclonal Antibody [clone: [3B10-5B10]]

Supplier: ProSci

51-479
10074-772EA 1124.21 CAD
10074-772
Anti-CK81 Mouse Monoclonal Antibody [clone: [3B10-5B10]]
Antibodies
KRT81 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, immunofluorescence, and immunoprecipitation.

PBS, pH 7.2

Store KRT81 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: KRT81
Clonality: Monoclonal
Clone: [3B10-5B10]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR