Anti-SMAD6 Mouse Monoclonal Antibody [clone: [1G2]]

Supplier: ProSci

51-484
10074-782EA 1124.21 CAD
10074-782
Anti-SMAD6 Mouse Monoclonal Antibody [clone: [1G2]]
Antibodies
SMAD6 antibody can be used in ELISA starting at 1:10000 - 1:50000, Western Blot starting at 1:200 - 1:2000, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

Store SMAD6 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: SMAD6
Clonality: Monoclonal
Clone: [1G2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR