Anti-MSI1 Mouse Monoclonal Antibody [clone: [3F2]]

Supplier: ProSci
51-487
10074-786EA 1123.86 CAD
10074-786
Anti-MSI1 Mouse Monoclonal Antibody [clone: [3F2]]
Antibodies
  • MSI1 antibody can be used in ELISA starting at 1:10000 - 1:50000, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 2.5 μg/mL.

Protein A Column

PBS, pH 7.2

Store MSI1 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: MSI1
Clonality: Monoclonal
Clone: [3F2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR