Anti-PLEK Mouse Monoclonal Antibody [clone: [6E3]]

Supplier: ProSci
51-497
10076-982EA 1124.21 CAD
10076-982
Anti-PLEK Mouse Monoclonal Antibody [clone: [6E3]]
Antibodies
PLEK antibody can be used in Western Blot, and immunohistochemistry starting at 1:500 - 1:1000.

PBS, pH 7.2

Store PLEK antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: PLEK
Clonality: Monoclonal
Clone: [6E3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR