Anti-TFF3 Mouse Monoclonal Antibody [clone: [3D9]]

51-519
10073-266EA 906.92 CAD
10073-266
Anti-TFF3 Mouse Monoclonal Antibody [clone: [3D9]]
TFF3 antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Store TFF3 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: TFF3
Clonality: Monoclonal
Clone: [3D9]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR