Anti-KHSRP Mouse Monoclonal Antibody [clone: [4C10]]

Supplier: ProSci

51-531
10077-034EA 1124.21 CAD
10077-034
Anti-KHSRP Mouse Monoclonal Antibody [clone: [4C10]]
Antibodies
KHSRP antibody can be used in Western Blot, and immunohistochemistry starting at 20 ?g/mL.

PBS, pH 7.2

Store KHSRP antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: KHSRP
Clonality: Monoclonal
Clone: [4C10]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR