Anti-GMPS Mouse Monoclonal Antibody [clone: [1D10]]

Supplier: ProSci

51-536
10077-044EA 1124.21 CAD
10077-044
Anti-GMPS Mouse Monoclonal Antibody [clone: [1D10]]
Antibodies
GMPS antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 1:400.

PBS, pH 7.2

Store GMPS antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: GMPS
Clonality: Monoclonal
Clone: [1D10]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR