Anti-CDC123 Mouse Monoclonal Antibody [clone: [4B9]]

Supplier: ProSci
51-537
10077-046EA 1201.59 CAD
10077-046
Anti-CDC123 Mouse Monoclonal Antibody [clone: [4B9]]
Antibodies
CDC123 antibody can be used in ELISA starting at 1: 10000, Western Blot starting at 1: 200 - 1: 1000, and immunohistochemistry starting at 1:200.

PBS, pH 7.2

Store CDC123 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: CDC123
Clonality: Monoclonal
Clone: [4B9]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR