Anti-STK17A Mouse Monoclonal Antibody [clone: [4D12]]

Supplier: ProSci

51-541
10077-054EA 1123.86 CAD
10077-054
Anti-STK17A Mouse Monoclonal Antibody [clone: [4D12]]
Antibodies
DRAK1 antibody can be used in Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

Store DRAK1 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: STK17A
Clonality: Monoclonal
Clone: [4D12]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR