Anti-SCGN Mouse Monoclonal Antibody [clone: [2G7]]

51-554
10077-080EA 906.92 CAD
10077-080
Anti-SCGN Mouse Monoclonal Antibody [clone: [2G7]]
SCGN antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Store SCGN antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: SCGN
Clonality: Monoclonal
Clone: [2G7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR