Anti-LST1 Mouse Monoclonal Antibody [clone: [LST1/02]]

Supplier: ProSci

51-561
10079-508EA 955.07 CAD
10079-508
Anti-LST1 Mouse Monoclonal Antibody [clone: [LST1/02]]
Antibodies
LST-1 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 20 ?g/mL, and flow cytometry starting at 0.1 µg/1E6 cells.

PBS, 15 mM sodium azide, pH 7.4

LST-1 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: LST1
Clonality: Monoclonal
Clone: [LST1/02]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR