Anti-SLC44A2 Mouse Monoclonal Antibody [clone: [3D11]]

Supplier: ProSci

51-588
10079-562EA 1124.21 CAD
10079-562
Anti-SLC44A2 Mouse Monoclonal Antibody [clone: [3D11]]
Antibodies
SLC44A2 antibody can be used in Western Blot, immunohistochemistry starting at 15 ?g/mL, and immunocytochemistry.

PBS, pH 7.2

Store SLC44A2 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: SLC44A2
Clonality: Monoclonal
Clone: [3D11]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR