Anti-HAMP Mouse Monoclonal Antibody [clone: [1F9]]

Supplier: ProSci
51-590
10079-566EA 1167.21 CAD
10079-566
Anti-HAMP Mouse Monoclonal Antibody [clone: [1F9]]
Antibodies
Hepcidin antibody can be used in Western Blot, immunohistochemistry starting at 1:50, and immunoprecipitation.

  • Storage buffer: PBS, pH 7.2

Store Hepcidin antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: HAMP
Clonality: Monoclonal
Clone: [1F9]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR