Anti-STK33 Mouse Monoclonal Antibody [clone: [3F10]]

Supplier: ProSci

51-591
10079-568EA 1124.21 CAD
10079-568
Anti-STK33 Mouse Monoclonal Antibody [clone: [3F10]]
Antibodies
STK33 antibody can be used in Western Blot, and immunohistochemistry starting at 1:50.

PBS, pH 7.2

Store STK33 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: STK33
Clonality: Monoclonal
Clone: [3F10]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR