Anti-EPSTI1 Mouse Monoclonal Antibody [clone: [2A8]]

Supplier: ProSci

51-599
10077-096EA 1124.21 CAD
10077-096
Anti-EPSTI1 Mouse Monoclonal Antibody [clone: [2A8]]
Antibodies
EPSTI1 antibody can be used in Western Blot, immunohistochemistry starting at 10 ?g/mL, and immunoprecipitation.

PBS, pH 7.2

Store EPSTI1 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: EPSTI1
Clonality: Monoclonal
Clone: [2A8]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR