To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:endothelial cell antibody
- Clonality:Monoclonal
- Clone:[HIS52]
- Gene ID:P48291
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat
- Antigen symbol:RECA-1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Form:Liquid
- Antigen synonyms:RECA-1
- Storage buffer:PBS, 0.09% sodium azide.
- Storage temperature:RECA-1 antibody can be stored short term 4 °C. For long term storage aliquot and store at –20 °C. As with all antibodies avoid freeze/thaw cycles.
- Concentration:0.5 mg/mL
- Shipping temperature:4 °C
- Immunogen:Stromal cells from rat lymph node
- Purification:Affinity Chromatography
- Cat. no.:10077-124
Specifications
About this item
RECA-1 antibody can be used in ELISA, Western Blot starting at 1:1000 - 1:2000, and immunohistochemistry starting at 5 ?g/mL.
PBS, 0.09% sodium azide.
RECA-1 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.
Type: Primary
Antigen: RECA-1
Clonality: Monoclonal
Clone: [HIS52]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat