Anti-Troponin T type 2 Mouse Monoclonal Antibody

51-628
10077-150EA 811.46 CAD
10077-150
Anti-Troponin T type 2 Mouse Monoclonal Antibody
TNNT2 antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.4, 0.1% sodium azide.

TNNT2 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: TNNT2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Bovine, Cat, Dog, Fish, Goat, Human, Mouse, Pig, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR