Anti-TG Mouse Monoclonal Antibody

Supplier: ProSci
51-647
10077-182EA 736.85 CAD
10077-182
Anti-TG Mouse Monoclonal Antibody
Thyroglobulin antibody can be used in Western Blot, and immunohistochemistry starting at 20 ?g/mL.

10 mM PBS, pH 7.4, BSA, 0.1% sodium azide.

Store Thyroglobulin antibody at 2 °C to 8 °C degrees. Do not freeze.

Type:
Antigen: TG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR