Anti-Troponin T type 2 Mouse Monoclonal Antibody [clone: [M322512]]

Supplier: ProSci
51-650
10073-276EA 811.46 CAD
10073-276
Anti-Troponin T type 2 Mouse Monoclonal Antibody [clone: [M322512]]
CTnT antibody can be used in Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunoprecipitation.

PBS buffer, pH 7.4 with 0.1% NaN3

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TNNT2
Clonality: Monoclonal
Clone: [M322512]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR