Anti-TUBB3 Mouse Monoclonal Antibody

Supplier: ProSci

51-652
10073-280EA 1011.8 CAD
10073-280
Anti-TUBB3 Mouse Monoclonal Antibody
Antibodies
Beta III Tubulin antibody can be used in Western Blot starting at 1:2000 - 1:5000, immunohistochemistry starting at 5 ?g/mL, and immunocytochemistry starting at 1:1000 - 1:2000.

0.1 M Tris-glycine, 0.15 M NaCl, pH 7.4, 0.05% sodium azide, 30% glycerol.

Beta III Tubulin antibody may be stored at 4 °C for short-term only. As with all antibodies avoid freeze/thaw cycles. Store at -20 °C for long term. Aliquots are stable for at least 12 months

Type: Primary
Antigen: TUBB3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Gibbon, Bovine, Dog, Gorilla, Human, Monkey, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR