Anti-RUNX1 Mouse Monoclonal Antibody [clone: [3A1]]

Supplier: ProSci
51-678
10074-816EA 906.92 CAD
10074-816
Anti-RUNX1 Mouse Monoclonal Antibody [clone: [3A1]]
RUNX1 antibody can be used in Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

RUNX1 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: RUNX1
Clonality: Monoclonal
Clone: [3A1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR