Anti-ARID3A Mouse Monoclonal Antibody [clone: [1A11]]

Supplier: ProSci

51-695
10073-314EA 1124.21 CAD
10073-314
Anti-ARID3A Mouse Monoclonal Antibody [clone: [1A11]]
Antibodies
ARID3A antibody can be used in Western Blot starting at 0.5 - 2 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

ARID3A antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: ARID3A
Clonality: Monoclonal
Clone: [1A11]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR