Anti-FGFR2 Mouse Monoclonal Antibody [clone: [1G3]]

Supplier: ProSci

51-704
10073-330EA 1061.16 CAD
10073-330
Anti-FGFR2 Mouse Monoclonal Antibody [clone: [1G3]]
Antibodies
FGFR2 antibody can be used in Western Blot starting at 1 - 3 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

FGFR2 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: FGFR2
Clonality: Monoclonal
Clone: [1G3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR