Anti-PCSK1 Mouse Monoclonal Antibody [clone: [3D2]]

Supplier: ProSci
51-707
10073-336EA 1167.21 CAD
10073-336
Anti-PCSK1 Mouse Monoclonal Antibody [clone: [3D2]]
Antibodies
PCSK1 antibody can be used in Western Blot starting at 0.5 - 2 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

PCSK1 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: PCSK1
Clonality: Monoclonal
Clone: [3D2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3,k
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR