Anti-GLUL Mouse Monoclonal Antibody

51-714
10073-350EA 757.17 CAD
10073-350
Anti-GLUL Mouse Monoclonal Antibody
Glutamine Synthetase antibody can be used in Western Blot starting at 0.5 - 1 ?g/mL, and immunohistochemistry starting at 5 - 10 ?g/mL.

PBS, pH 7.2

Glutamine Synthetase antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type:
Antigen: GLUL
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR