Anti-TRIM28 Mouse Monoclonal Antibody [clone: [4E6]]

Supplier: ProSci

51-735
10077-208EA 1124.21 CAD
10077-208
Anti-TRIM28 Mouse Monoclonal Antibody [clone: [4E6]]
Antibodies
TRIM28 antibody can be used in Western Blot starting at 1 - 3 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

TRIM28 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: TRIM28
Clonality: Monoclonal
Clone: [4E6]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR