Anti-TRIM28 Mouse Monoclonal Antibody [clone: [1D11]]

Supplier: ProSci

51-736
10077-210EA 1124.21 CAD
10077-210
Anti-TRIM28 Mouse Monoclonal Antibody [clone: [1D11]]
Antibodies
TRIM28 antibody can be used in Western Blot starting at 0.5 - 4 ?g/mL, and immunohistochemistry starting at 20 ?g/mL.

PBS, pH 7.2

TRIM28 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: TRIM28
Clonality: Monoclonal
Clone: [1D11]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR