Anti-MEOX2 Mouse Monoclonal Antibody [clone: [6A5]]

Supplier: ProSci

51-765
10074-860EA 1124.21 CAD
10074-860
Anti-MEOX2 Mouse Monoclonal Antibody [clone: [6A5]]
Antibodies
MEOX2 antibody can be used in ELISA starting at 1 ?g/mL, Western Blot starting at 1 ?g/mL, and immunohistochemistry starting at 20 ?g/mL.

PBS, pH 7.2

MEOX2 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: MEOX2
Clonality: Monoclonal
Clone: [6A5]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR