Anti-MRC1 Mouse Monoclonal Antibody

Supplier: ProSci
51-767
10074-864EA 884.22 CAD
10074-864
Anti-MRC1 Mouse Monoclonal Antibody
CD206 antibody can be used in Western Blot starting at 0.5 - 2 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

CD206 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type:
Antigen: MRC1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR