Anti-NEK2 Mouse Monoclonal Antibody [clone: [2F6]]

Supplier: ProSci

51-771
10074-872EA 1124.21 CAD
10074-872
Anti-NEK2 Mouse Monoclonal Antibody [clone: [2F6]]
Antibodies
NEK2 antibody can be used in Western Blot, immunohistochemistry starting at 10 ?g/mL, immunoprecipitation, and flow cytometry.

PBS, pH 7.2

NEK2 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: NEK2
Clonality: Monoclonal
Clone: [2F6]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR