Anti-PSAP Mouse Monoclonal Antibody [clone: [1D1-C12]]

Supplier: ProSci
51-791
10077-256EA 906.92 CAD
10077-256
Anti-PSAP Mouse Monoclonal Antibody [clone: [1D1-C12]]
PSAP antibody can be used in Western Blot starting at 1 - 2 ?g/mL, immunohistochemistry starting at 5 ?g/mL, and immunocytochemistry.

PBS, pH 7.2

PSAP antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: PSAP
Clonality: Monoclonal
Clone: [1D1-C12]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR