Anti-SGK1 Mouse Monoclonal Antibody [clone: [3E3]]

Supplier: ProSci

51-801
10073-372EA 1061.16 CAD
10073-372
Anti-SGK1 Mouse Monoclonal Antibody [clone: [3E3]]
Antibodies
SGK1 antibody can be used in Western Blot starting at 2 ?g/mL, immunohistochemistry starting at 5 ?g/mL, immunocytochemistry, and immunoprecipitation

PBS, pH 7.2

SGK1 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: SGK1
Clonality: Monoclonal
Clone: [3E3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR