Anti-SYT1 Mouse Monoclonal Antibody [clone: [1A6]]

Supplier: ProSci

51-812
10074-922EA 1124.21 CAD
10074-922
Anti-SYT1 Mouse Monoclonal Antibody [clone: [1A6]]
Antibodies
Synaptotagmin antibody can be used in Western Blot starting at 1:250 - 1:500, and immunohistochemistry starting at 1:50.

PBS, pH 7.2

Synaptotagmin antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: SYT1
Clonality: Monoclonal
Clone: [1A6]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR