Anti-CK7 Mouse Monoclonal Antibody [clone: [OV-TL 12/30]]

Supplier: ProSci
51-823
10079-600EA 811.46 CAD
10079-600
Anti-CK7 Mouse Monoclonal Antibody [clone: [OV-TL 12/30]]
KRT7 antibody can be used in ELISA, Western Blot starting at 1:1000 - 1:2000, and immunohistochemistry starting at 10 ?g/mL.

PBS, 0.1% BSA, 0.1% sodium azide

Store KRT7 antibody at 4 °C.

Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: [OV-TL 12/30]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR