Anti-SELPLG Rat Monoclonal Antibody [clone: [HECA-452]]

Supplier: ProSci

51-833
10079-618EA 1011.8 CAD
10079-618
Anti-SELPLG Rat Monoclonal Antibody [clone: [HECA-452]]
Antibodies
SELPLG antibody can be used in immunohistochemistry starting at 10 ?g/mL.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide.

Aliquot and store at 4ºC. Avoid freezing.

Type: Primary
Antigen: SELPLG
Clonality: Monoclonal
Clone: [HECA-452]
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR