Anti-CD44 Rat Monoclonal Antibody [clone: [IM7]]

Supplier: ProSci
51-846
10073-394EA 811.46 CAD
10073-394
Anti-CD44 Rat Monoclonal Antibody [clone: [IM7]]
CD44 antibody can be used in immunohistochemistry starting at 10 ?g/mL.

PBS, 15 mM sodium azide, pH 7.4

CD44 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: [IM7]
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Cat, Dog, Horse, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR