Anti-PF4 Sheep Polyclonal Antibody

Supplier: ProSci
51-849
10073-398EA 1047.24 CAD
10073-398
Anti-PF4 Sheep Polyclonal Antibody
Antibodies
PF4 antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

0.01 M HEPES, pH 7.4, 0.15 M NaCl, 50% glycerol. No preservative added.

Store PF4 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: PF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR