Anti-MUC1 Hamster Monoclonal Antibody

51-858
10073-416EA 620.52 CAD
10073-416
Anti-MUC1 Hamster Monoclonal Antibody
MUC1 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 1:25.

10mM PBS, pH 7.4, with 0.2% BSA and 0.09% Sodium Azide

MUC1 Monoclonal Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MUC1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Gibbon, Gorilla, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR