Anti-AHR Mouse Monoclonal Antibody [clone: [3B9]]

Supplier: ProSci

51-879
10073-450EA 1124.21 CAD
10073-450
Anti-AHR Mouse Monoclonal Antibody [clone: [3B9]]
Antibodies
AHR antibody can be used in immunohistochemistry starting at 5 ?g/mL, and flow cytometry.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AHR
Clonality: Monoclonal
Clone: [3B9]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR