Anti-ARNT Mouse Monoclonal Antibody [clone: [3D10]]

Supplier: ProSci
51-884
10073-460EA 1123.86 CAD
10073-460
Anti-ARNT Mouse Monoclonal Antibody [clone: [3D10]]
Antibodies
ARNT antibody can be used in Western Blot starting at 0.5 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ARNT
Clonality: Monoclonal
Clone: [3D10]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR