Anti-BIK Mouse Monoclonal Antibody [clone: [4G12]]

Supplier: ProSci
51-888
10079-628EA 1124.21 CAD
10079-628
Anti-BIK Mouse Monoclonal Antibody [clone: [4G12]]
Antibodies
BIK antibody can be used in Western Blot starting at 2 ?g/mL, and immunohistochemistry starting at 1:50.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BIK
Clonality: Monoclonal
Clone: [4G12]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR