Anti-CAMK4 Mouse Monoclonal Antibody [clone: [1A3]]

Supplier: ProSci

51-890
10079-632EA 1124.21 CAD
10079-632
Anti-CAMK4 Mouse Monoclonal Antibody [clone: [1A3]]
Antibodies
CAMK4 antibody can be used in Western Blot starting at 2 ?g/mL, and immunohistochemistry starting at 1:25.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CAMK4
Clonality: Monoclonal
Clone: [1A3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR