Anti-CANX Mouse Monoclonal Antibody [clone: [1D4]]

Supplier: ProSci
51-891
10079-634EA 906.92 CAD
10079-634
Anti-CANX Mouse Monoclonal Antibody [clone: [1D4]]
Calnexin antibody can be used in immunohistochemistry starting at 10 ?g/mL, and flow cytometry starting at 1.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CANX
Clonality: Monoclonal
Clone: [1D4]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR