Anti-CD247 Mouse Monoclonal Antibody [clone: [4A12-F6]]

Supplier: ProSci

51-898
10073-464EA 1123.86 CAD
10073-464
Anti-CD247 Mouse Monoclonal Antibody [clone: [4A12-F6]]
Antibodies
CD3-Zeta antibody can be used in immunohistochemistry starting at 10 ?g/mL, and flow cytometry starting at 1 µg/1E6 cells.

PBS, pH 7.2

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CD247
Clonality: Monoclonal
Clone: [4A12-F6]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR