Anti-KIT Mouse Monoclonal Antibody [clone: [4F7]]

Supplier: Prosci

51-922
10073-504EA 1124.21 CAD
10073-504
Anti-KIT Mouse Monoclonal Antibody [clone: [4F7]]
Antibodies
KIT antibody can be used in Western Blot starting at 1 - 2 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

KIT antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: KIT
Clonality: Monoclonal
Clone: [4F7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR